Cyclobutyl phenyl ketone ≥95.0%
Supplier: TCI America
Synonyms:
Cyclobutyl Phenyl Ketone
C1035-5G
TCC1035-005GEA
70.24
USD
TCC1035-005G
Cyclobutyl phenyl ketone ≥95.0%
Cyclobutyl phenyl ketone
CAS Number: 5407-98-7
MDL Number: MFCD00001326
Molecular Formula: C11H12O
Molecular Weight: 160.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Benzoylcyclobutane
Formula:
C₁₁H₁₂O MW: 160.2124 g/mol Boiling Pt: 115 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001326 CAS Number: 5407-98-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...