3-(4-Carboxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-(2-Carboxyethyl)benzoic Acid
C1023-5G
TCC1023-005GEA
262
USD
TCC1023-005G
3-(4-Carboxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)
3-(4-Carboxyphenyl)propionic acid
CAS Number: 38628-51-2
MDL Number: MFCD00016560
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 4-(2-Carboxyethyl)benzoic Acid
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol |
MDL Number:
MFCD00016560 CAS Number: 38628-51-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...