(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98.0% (by titrimetric analysis)

Synonyms: L-Cysteine ethyl ester hydrochloride

C0996-25G
TCC0996-025GEA 36.95 USD
TCC0996-025G
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride

CAS Number: 868-59-7
MDL Number: MFCD00012631
Molecular Formula: C5H11NO2S
Molecular Weight: 185.67
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 128
Specific rotation [a]20/D: -11.5 deg (C=8, 1mol/L HCl)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Cys-OEt.HCl

Formula: C5H12ClNO2S
MW: 185.666 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00012631
CAS Number: 868-59-7
https://us.vwr.com/stibo/web/std.lang.all/82/60/28938260.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR