Cyclopropyldiphenylcarbinol ≥98.0%

Supplier: TCI America

Synonyms: Cyclopropyldiphenylmethanol

C0983-5G
TCC0983-5GEA 111.38 USD
TCC0983-5G
Cyclopropyldiphenylcarbinol ≥98.0%
Cyclopropyldiphenylcarbinol

CAS Number: 5785-66-0
MDL Number: MFCD00001298
Molecular Formula: C16H16O
Molecular Weight: 224.30
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 85


Other Product Names / Synonyms: Cyclopropyldiphenylmethanol

Formula: C₁₆H₁₆O
Boiling Pt: 150 °C
Melting Pt: 85 °C
MDL Number: MFCD00001298
CAS Number: 5785-66-0
https://us.vwr.com/stibo/web/std.lang.all/82/35/28938235.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR