1-Cyclohexyl-2-pyrrolidone ≥99.0%
Supplier: TCI America
C0963-25ML
C0963-500ML
TCC0963-500MLEA
94.1
USD
TCC0963-500ML
TCC0963-25ML
1-Cyclohexyl-2-pyrrolidone ≥99.0%
1-Cyclohexyl-2-pyrrolidone
CAS Number: 6837-24-7
MDL Number: MFCD00003191
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 141
Freezing point (°C): 15
Specific Gravity (20/20): 1.03
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Boiling Pt: 154 °C Density: 1.03 (20 °C) Flash Pt: 141 °C |
MDL Number:
MFCD00003191 CAS Number: 6837-24-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...