Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Chlorophenoxy)isobutyric acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Clofibric acid2-(4-Chlorophenoxy)-2-methylpropionic acid

C0940-25G C0940-500G
TCC0940-025G TCC0940-500G
2-(4-Chlorophenoxy)isobutyric acid ≥97.0% (by titrimetric analysis)
CAS Number: 882-09-7
MDL Number: MFCD00004192
Molecular Formula: C10H11ClO3
Molecular Weight: 214.65
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 120

Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: Clofibric Acid, PCIB


Formula: C₁₀H₁₁ClO₃
MW: 214,65
Melting Pt: 120 °C
MDL Number: MFCD00004192
CAS Number: 882-09-7
EINECS: 212-925-9
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)
Melting point (°C) 120


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty