Citronellyl acetate ≥95.0%

Supplier: TCI America

Synonyms: 3,7-Dimethyl-6-octen-1-ol Acetate

C0915-25ML
TCC0915-025MLEA 76 USD
TCC0915-025ML
Citronellyl acetate ≥95.0%
Citronellyl acetate

CAS Number: 150-84-5
MDL Number: MFCD00015039
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 112
Specific Gravity (20/20): 0.89


Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-6-octen-1-ol Acetate

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 173 °C
Density: 0.89 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00015039
CAS Number: 150-84-5
https://us.vwr.com/stibo/web/std.lang.all/81/33/28938133.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR