Cyclohexanecarboxaldehyde ≥98.0%
Supplier: TCI America
C0880-25ML
C0880-100ML
TCC0880-100MLEA
186.7
USD
TCC0880-100ML
TCC0880-025ML
Cyclohexanecarboxaldehyde ≥98.0%
Cyclohexanecarboxaldehyde
CAS Number: 2043-61-0
MDL Number: MFCD00001457
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 159
Flash Point (°C): 40
Specific Gravity (20/20): 0.93
Formula:
C₇H₁₂O Boiling Pt: 159 °C Density: 0.93 (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00001457 CAS Number: 2043-61-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...