S-(Carboxymethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Carbocysteine3-[(Carboxymethyl)thio]alanine

C0863-25G
TCC0863-025GEA 36.71 USD
TCC0863-025G
S-(Carboxymethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
S-(Carboxymethyl)-L-cysteine

CAS Number: 638-23-3
MDL Number: MFCD00002614
Molecular Formula: C5H9NO4S
Molecular Weight: 179.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: Carbocysteine, 3-[(Carboxymethyl)thio]alanine

Formula: C₅H₉NO₄S
MW: 179.19 g/mol
MDL Number: MFCD00002614
CAS Number: 638-23-3
https://us.vwr.com/stibo/web/std.lang.all/80/07/28938007.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR