1-(Cyanomethyl)pyridinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(Cyanomethyl)pyridin-1-ium chloride

C0848-5G
TCC0848-005GEA 55 USD
TCC0848-005G
1-(Cyanomethyl)pyridinium chloride ≥98.0% (by titrimetric analysis)
1-(Cyanomethyl)pyridinium chloride

CAS Number: 17281-59-3
MDL Number: MFCD00031933
Molecular Formula: C7H7ClN2
Molecular Weight: 154.60
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 174

Formula: C₇H₇ClN₂
MW: 154.60 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00031933
CAS Number: 17281-59-3
https://us.vwr.com/stibo/web/std.lang.all/79/91/28937991.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR