2-Cyclopentylethanol ≥98.0%

Supplier: TCI America

C0817-5ML C0817-25ML
TCC0817-005MLEA 207.06 USD
TCC0817-005ML TCC0817-25ML
2-Cyclopentylethanol ≥98.0%
2-Cyclopentylethanol

CAS Number: 766-00-7
MDL Number: MFCD00019301
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 70
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 2-Cyclopentylethanol, 2-Hydroxyethylcyclopentane

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 182 °C
Density: 0.92 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00019301
CAS Number: 766-00-7
https://us.vwr.com/stibo/web/std.lang.all/78/56/28937856.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR