ɑ-Cyclopentylphenylacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Cyclopentyl-2-phenylacetic acid
, ɑ-Phenylcyclopentaneacetic Acid
C0808-10G
TCC0808-010GEA
82.47
USD
TCC0808-010G
ɑ-Cyclopentylphenylacetic acid ≥98.0% (by titrimetric analysis)
alpha-Cyclopentylphenylacetic acid
CAS Number: 3900-93-4
MDL Number: MFCD00001379
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 100
Other Product Names / Synonyms: alpha-Phenylcyclopentaneacetic Acid
Formula:
C₁₃H₁₆O₂ MW: 204.27 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD00001379 CAS Number: 3900-93-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...