TCIAC0725

C0725-10G
TCC0725-10GEA 305.86 USD
TCC0725-10G
TCIAC0725
CAS Number: 15455-80-8
MDL Number: MFCD00059504
Molecular Formula: C34H58O3
Molecular Weight: 514.84
Form: Crystal
Color: White
Melting point (°C): 107
Specific rotation [a]20/D: -31 deg (C=2, CHCl3)

Other Product Names / Synonyms: Hexyl Carbonic Acid Cholesterol Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR