TCIAC0721

C0721-10G
TCC0721-10GEA 305.86 USD
TCC0721-10G
TCIAC0721
CAS Number: 15455-82-0
MDL Number: MFCD00059873
Molecular Formula: C36H62O3
Molecular Weight: 542.89
Form: Crystal
Color: White

Other Product Names / Synonyms: n-Octyl Carbonic Acid Cholesterol Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR