TCIAC0620

C0620-25G
TCC0620-25GEA 175.71 USD
TCC0620-25G
TCIAC0620
CAS Number: 1908-11-8
MDL Number: MFCD00021142
Molecular Formula: C39H68O2
Molecular Weight: 568.97
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 91
Specific rotation [a]20/D: -22 deg (C=2, THF)

Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials), Bioscience, Lipids
Other Product Names / Synonyms: Cholesterol Dodecanoate, Lauric Acid Cholesterol Ester, Dodecanoic Acid Cholesterol Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR