Crocein scarlet 3B

Supplier: TCI America

Synonyms: Acid Red 73

C0531-25G
TCC0531-025GEA 33.83 USD
TCC0531-025G
Crocein scarlet 3B
Crocein scarlet 3B

CAS Number: 5413-75-2
MDL Number: MFCD00003907
Molecular Formula: C22H16N4O7S2
Molecular Weight: 556.47
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Acid Red 73

Formula: C₂₂H₁₆N₄O₇S₂
MDL Number: MFCD00003907
CAS Number: 5413-75-2
https://us.vwr.com/stibo/web/std.lang.all/75/61/28937561.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR