TCIAC0319

C0319-500G C0319-25G
TCC0319-500GEA 421.82 USD
TCC0319-500G TCC0319-25G
TCIAC0319
CAS Number: 604-35-3
MDL Number: MFCD00003636
Molecular Formula: C29H48O2
Molecular Weight: 428.70
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 113
Specific rotation [a]20/D: -40 deg (C=2, Toluene)

Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials), Bioscience, Lipids
Other Product Names / Synonyms: Acetic Acid Cholesterol Ester, Acetyl Cholesterol
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR