8-Chlorotheophylline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C0293-250G
C0293-25G
TCC0293-025GEA
29.02
USD
TCC0293-025G
TCC0293-250G
8-Chlorotheophylline ≥98.0% (by HPLC, titration analysis)
8-Chlorotheophylline
CAS Number: 85-18-7
MDL Number: MFCD00005581
Molecular Formula: C7H7ClN4O2
Molecular Weight: 214.61
Purity/Analysis Method:
Form: Crystal
Formula:
C₇H₇ClN₄O₂ |
MDL Number:
MFCD00005581 CAS Number: 85-18-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...