2-tert-Butylpyrazine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3394-5G B3394-1G
TCB3394-5GEA 327.45 USD
TCB3394-5G TCB3394-1G
2-tert-Butylpyrazine ≥97.0% (by GC, titration analysis)
2-tert-Butylpyrazine

CAS Number: 32741-11-0
MDL Number: MFCD00069193
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.96

Formula: C₈H₁₂N₂
Density: 0.96 (20 °C)
MDL Number: MFCD00069193
CAS Number: 32741-11-0
https://us.vwr.com/stibo/web/std.lang.all/47/74/28934774.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR