3-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
Synonyms:
1-Bromo-2-fluoro-3-(trifluoromethyl)benzene
, 3-Bromo-2-fluorobenzotrifluoride
CAS Number: 144584-67-8
MDL Number: MFCD00070812
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 169
Flash Point (°C): 100
Specific Gravity (20/20): 1.74
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Bromo-2-fluoro-1-(trifluoromethyl)benzene, 3-Bromo-alpha,alpha,alpha,2-tetrafluorotoluene
Formula:
C₇H₃BrF₄ MW: 243 g/mol Boiling Pt: 169 °C Density: 1.74 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00070812 CAS Number: 144584-67-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...