Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0%

Supplier: TCI America

B3388-1G B3388-5G
TCB3388-1G TCB3388-5G
2-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0%
CAS Number: 104540-42-3
MDL Number: MFCD00061243
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 88
Specific Gravity (20/20): 1.76

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-3-fluoro-1-(trifluoromethyl)benzene, 2-Bromo-alpha,alpha,alpha,3-tetrafluorotoluene


Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 168 °C
Density: 1,76 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00061243
CAS Number: 104540-42-3
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)
Boiling point (°C) 168
Flash Point (°C) 88
Specific Gravity (20/20) 1.76
Storage Temperature 0-10°C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty