2-Bromoisonicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Bromo-4-pyridinecarboxylic acid
, 2-Bromopyridine-4-carboxylic acid
B3368-1G
B3368-5G
TCB3368-1GEA
33
USD
TCB3368-1G
TCB3368-5G
2-Bromoisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Bromoisonicotinic acid
CAS Number: 66572-56-3
MDL Number: MFCD01646069
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 229
Other Product Names / Synonyms: 2-Bromo-4-pyridinecarboxylic Acid
Formula:
C₆H₄BrNO₂ Melting Pt: 229 °C |
MDL Number:
MFCD01646069 CAS Number: 66572-56-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...