5-Bromoisoquinoline ≥96.0% (by GC, titration analysis)

Supplier: TCI America

B3313-1G B3313-5G
TCB3313-1GEA 38 USD
TCB3313-1G TCB3313-5G
5-Bromoisoquinoline ≥96.0% (by GC, titration analysis)
5-Bromoisoquinoline

CAS Number: 34784-04-8
MDL Number: MFCD01646405
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: 96.0% (GC,T)
Form: Crystal
Boiling point (°C): 97
Melting point (°C): 82

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 97 °C
Melting Pt: 82 °C
MDL Number: MFCD01646405
CAS Number: 34784-04-8
https://us.vwr.com/stibo/web/std.lang.all/47/05/28934705.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR