4'-Bromo-3'-methylacetanilide ≥98.0%
Supplier: TCI America
B3142-5G
B3142-25G
TCB3142-25GEA
87
USD
TCB3142-25G
TCB3142-5G
4'-Bromo-3'-methylacetanilide ≥98.0%
4'-Bromo-3'-methylacetanilide
CAS Number: 90914-81-1
MDL Number: MFCD00040822
Molecular Formula: C9H10BrNO
Molecular Weight: 228.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 103
Other Product Names / Synonyms: N-Acetyl-4-bromo-m-toluidine
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...