4'-Bromo-3'-chloroacetanilide ≥98.0%

Supplier: TCI America

B3141-25G B3141-5G
TCB3141-25GEA 161 USD
TCB3141-25G TCB3141-5G
4'-Bromo-3'-chloroacetanilide ≥98.0%
4'-Bromo-3'-chloroacetanilide

CAS Number: 22459-81-0
MDL Number: MFCD00040848
Molecular Formula: C8H7BrClNO
Molecular Weight: 248.50
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 125

Formula: H₃CC(O)NH(C₆H₃BrCl)
Melting Pt: 125 °C
MDL Number: MFCD00040848
CAS Number: 22459-81-0
https://us.vwr.com/stibo/web/std.lang.all/45/76/28934576.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR