2-Bromobenzaldehyde oxime ≥98.0%
Supplier: TCI America
Synonyms:
2-Bromobenzaldoxime
B3140-5G
TCB3140-5GEA
80.15
USD
TCB3140-5G
2-Bromobenzaldehyde oxime ≥98.0%
2-Bromobenzaldehyde oxime
CAS Number: 34158-72-0
MDL Number: MFCD00082734
Molecular Formula: C7H6BrNO
Molecular Weight: 200.04
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 103
Formula:
C₇H₆BrNO MW: 200.03 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00082734 CAS Number: 34158-72-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...