Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Bromoacetanilide ≥98.0%

Supplier: TCI America


B3139-25G B3139-5G
TCB3139-25GEA 261.15 USD InStock
TCB3139-25G TCB3139-5G
2'-Bromoacetanilide ≥98.0%
2'-Bromoacetanilide
CAS Number: 614-76-6
MDL Number: MFCD00099252
Molecular Formula: C8H8BrNO
Molecular Weight: 214.06
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 102


Formula: H₃CC(O)NH(C₆H₄Br)
MW: 214.06 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00099252
CAS Number: 614-76-6
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty