3-(4-Bromobenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-(4-Bromophenyl)-4-oxobutanoic acid
B3129-5G
B3129-25G
TCB3129-25GEA
200.85
USD
TCB3129-25G
TCB3129-5G
3-(4-Bromobenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-(4-Bromobenzoyl)propionic acid
CAS Number: 6340-79-0
MDL Number: MFCD00016563
Molecular Formula: C10H9BrO3
Molecular Weight: 257.08
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 150
Formula:
C₁₀H₉BrO₃ MW: 257.08 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00016563 CAS Number: 6340-79-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...