5-Benzyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3086-25G
TCB3086-25GEA 125 USD
TCB3086-25G
5-Benzyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-Benzyl-1H-tetrazole

CAS Number: 18489-25-3
MDL Number: MFCD00068730
Molecular Formula: C8H8N4
Molecular Weight: 160.18
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 127

Formula: C₈H₈N₄
MW: 160,18
Melting Pt: 127 °C
MDL Number: MFCD00068730
CAS Number: 18489-25-3
EINECS: 000-000-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR