2-Bromofluorene ≥98.0%

Supplier: TCI America

B3069-25G B3069-100G B3069-5G
TCB3069-100GEA 262 USD
TCB3069-100G TCB3069-5G TCB3069-25G
2-Bromofluorene ≥98.0%
2-Bromofluorene

CAS Number: 1133-80-8
MDL Number: MFCD00001115
Molecular Formula: C13H9Br
Molecular Weight: 245.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 114

Formula: C₁₃H₉Br
MW: 245.12 g/mol
Boiling Pt: 185 °C
Melting Pt: 114 °C
MDL Number: MFCD00001115
CAS Number: 1133-80-8
https://us.vwr.com/stibo/web/std.lang.all/45/09/28934509.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR