Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-fluorobenzonitrile ≥97.0%

Supplier: TCI America


B3010-25G B3010-5G
TCB3010-25GEA 155.03 USD InStock
TCB3010-25G TCB3010-5G
2-Bromo-5-fluorobenzonitrile ≥97.0%
2-Bromo-5-fluorobenzonitrile
CAS Number: 57381-39-2
MDL Number: MFCD00142875
Molecular Formula: C7H3BrFN
Molecular Weight: 200.01
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 90


Formula: C₇H₃BrFN
MW: 200,01 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 90 °C
Storage Temperature: ambient
MDL Number: MFCD00142875
CAS Number: 57381-39-2
EINECS: 260-711-9
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty