Benzo[b]furan-2-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
Benzofuran-2-boronic acid
B2978-1G
TCB2978-1GEA
42.26
USD
TCB2978-1G
Benzo[b]furan-2-boronic acid (contains varying amounts of Anhydride)
Benzo[b]furan-2-boronic acid
CAS Number: 98437-24-2
MDL Number: MFCD00236019
Molecular Formula: C8H7BO3
Molecular Weight: 161.95
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 135
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...