Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromonicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Bromopyridine-3-carboxylic acid2-Bromo-5-pyridinecarboxylic acid6-Bromo-3-pyridinecarboxylic acid

B2961-5G B2961-25G
TCB2961-5GEA 116.52 USD InStock
TCB2961-5G TCB2961-25G
6-Bromonicotinic acid ≥98.0% (by GC, titration analysis)
6-Bromonicotinic acid
CAS Number: 6311-35-9
MDL Number: MFCD01927100
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 190

Other Product Names / Synonyms: 6-Bromopyridine-3-carboxylic Acid


Formula: C₆H₄BrNO₂
MW: 202,01 g/mol
Melting Pt: 190 °C
MDL Number: MFCD01927100
CAS Number: 6311-35-9
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty