2-Bromophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
Dihydroxy(2-bromophenyl)borane
, (2-Bromophenyl)metaboric acid
, (2-Bromophenyl)boronic acid
, 2-Bromobenzeneboronic acid
B2889-5G
TCB2889-5GEA
64.96
USD
TCB2889-5G
2-Bromophenylboronic acid (contains varying amounts of Anhydride)
2-Bromophenylboronic acid
CAS Number: 244205-40-1
MDL Number: MFCD01114672
Molecular Formula: C6H6BBrO2
Molecular Weight: 200.83
Form: Crystal
Color: White
Melting point (°C): 110
Other Product Names / Synonyms: 2-Bromobenzeneboronic Acid
Formula:
C₆H₆BBrO₂ MW: 200.83 g/mol Melting Pt: 110 °C |
MDL Number:
MFCD01114672 CAS Number: 244205-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...