4-tert-Butylbenzhydrazide ≥98.0%
Supplier: TCI America
Synonyms:
4-tert-Butylbenzoic hydrazide
, 4-tert-Butylbenzene-1-carbohydrazide
, 4-tert-Butylbenzohydrazide
, 4-tert-Butylbenzoic acid hydrazide
, 4-(tert-Butyl)benzohydrazide
CAS Number: 43100-38-5
MDL Number: MFCD00014763
Molecular Formula: C11H16N2O
Molecular Weight: 192.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 130
Other Product Names / Synonyms: 4-tert-Butylbenzoic Hydrazide, 4-tert-Butylbenzoylhydrazine
Formula:
C₁₁H₁₆N₂O MW: 192.26 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00014763 CAS Number: 43100-38-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...