4-Bromobenzaldehyde diethyl acetal ≥97.0%
Supplier: TCI America
Synonyms:
4-Bromo-α,α-diethoxytoluene
, 1-Bromo-4-(diethoxymethyl)benzene
B2845-25G
TCB2845-25GEA
61.92
USD
TCB2845-25G
4-Bromobenzaldehyde diethyl acetal ≥97.0%
4-Bromobenzaldehyde diethyl acetal
CAS Number: 34421-94-8
Molecular Formula: C11H15BrO2
Molecular Weight: 259.14
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 105
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: 1-Bromo-4-(diethoxymethyl)benzene
Formula:
C₁₁H₁₅BrO₂ MW: 259.14 g/mol Density: 1.28 (20 °C) Flash Pt: 105 °C |
MDL Number:
MFCD01863514 CAS Number: 34421-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...