1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥98.0%
Synonyms:
1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole
B2636-5G
TCB2636-5GEA
101.97
USD
TCB2636-5G
1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥98.0%
1-(4-Bromophenyl)-2,5-dimethylpyrrole
CAS Number: 5044-24-6
Molecular Formula: C12H12BrN
Molecular Weight: 250.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 74
Formula:
C₁₂H₁₂BrN MW: 250.14 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD00277042 CAS Number: 5044-24-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...