4'-Bromo-2'-chloroacetanilide ≥98.0%

Supplier: TCI America

B2413-5G B2413-25G
TCB2413-5GEA 34.83 USD InStock
TCB2413-5G TCB2413-25G
4'-Bromo-2'-chloroacetanilide ≥98.0%
4'-Bromo-2'-chloroacetanilide

CAS Number: 3460-23-9
MDL Number: MFCD00040852
Molecular Formula: C8H7BrClNO
Molecular Weight: 248.50
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 151

Formula: H₃CC(O)NH(C₆H₃BrCl)
MW: 248,51 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00040852
CAS Number: 3460-23-9
EINECS: 000-000-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR