Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Benzyloxy)benzaldehyde ≥98.0%

Supplier: TCI America


B2400-25G
TCB2400-25GEA 58.27 USD InStock
TCB2400-25G
3-(Benzyloxy)benzaldehyde ≥98.0%
3-(Benzyloxy)benzaldehyde
CAS Number: 1700-37-4
MDL Number: MFCD00003367
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 57


Formula: C₁₄H₁₂O₂
MW: 212,25 g/mol
Boiling Pt: 119 to 120 °C (0.02 torr)
Melting Pt: 57 °C
Storage Temperature: ambient
MDL Number: MFCD00003367
CAS Number: 1700-37-4
EINECS: 216-932-8
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty