4-Bromo-2-nitrophenol ≥98.0%

Supplier: TCI America

B2363-5G B2363-25G
TCB2363-5GEA 35.53 USD
TCB2363-5G TCB2363-25G
4-Bromo-2-nitrophenol ≥98.0%
4-Bromo-2-nitrophenol

CAS Number: 7693-52-9
MDL Number: MFCD00082540
Molecular Formula: C6H4BrNO3
Molecular Weight: 218.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 90


Other Product Names / Synonyms: 4-Bromo-1-hydroxy-2-nitrobenzene

Formula: C₆H₄BrNO₃
Melting Pt: 90 °C
MDL Number: MFCD00082540
CAS Number: 7693-52-9
https://us.vwr.com/stibo/web/std.lang.all/41/10/28934110.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR