2-Bromostyrene ≥95.0% stabilized

Supplier: TCI America

B2355-5G
TCB2355-5GEA 277 USD
TCB2355-5G
2-Bromostyrene ≥95.0% stabilized
2-Bromostyrene

(stabilized with TBC)
CAS Number: 2039-88-5
MDL Number: MFCD00000076
Molecular Formula: C8H7Br
Molecular Weight: 183.05
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Melting point (°C): -53
Flash Point (°C): 86
Specific Gravity (20/20): 1.41
Storage Temperature: 0-10°C

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 61 °C
Melting Pt: –53 °C
Density: 1,41 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00000076
CAS Number: 2039-88-5

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR