6-Bromo-1,4-benzodioxane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Bromo-2,3-dihydro-1,4-benzodioxine6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

B2351-5G
TCB2351-5GEA 68 USD
TCB2351-5G
6-Bromo-1,4-benzodioxane ≥98.0% (by titrimetric analysis)
6-Bromo-1,4-benzodioxane

CAS Number: 52287-51-1
MDL Number: MFCD00040750
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.61


Other Product Names / Synonyms: 4-Bromo-1,2-ethylenedioxybenzene

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Density: 1.61 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00040750
CAS Number: 52287-51-1
https://us.vwr.com/stibo/web/std.lang.all/41/01/28934101.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR