3-Bromo-4-hydroxybenzaldehyde ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B2350-5G
TCB2350-5GEA
66.99
USD
TCB2350-5G
3-Bromo-4-hydroxybenzaldehyde ≥98.0% (by titrimetric analysis)
3-Bromo-4-hydroxybenzaldehyde
CAS Number: 2973-78-6
MDL Number: MFCD00017348
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 126
Formula:
C₇H₅BrO₂ Melting Pt: 126 °C |
MDL Number:
MFCD00017348 CAS Number: 2973-78-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...