Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromomethyl-1,4-benzodioxane ≥98.0% (by GC, titration analysis)

Supplier: TCI America


B2349-1G B2349-5G
TCB2349-1GEA 33.29 USD InStock
TCB2349-1G TCB2349-5G
2-Bromomethyl-1,4-benzodioxane ≥98.0% (by GC, titration analysis)
2-Bromomethyl-1,4-benzodioxane
CAS Number: 2164-34-3
MDL Number: MFCD00044831
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.54


Formula: C₉H₉BrO₂
MW: 229,07 g/mol
Boiling Pt: 63 to 64 °C (0.005 torr)
Density: 1,54 (20 °C)
MDL Number: MFCD00044831
CAS Number: 2164-34-3
EINECS: 218-503-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty