Benzothiazol-2-ol ≥98.0%
Supplier: TCI America
Synonyms:
2-Hydroxybenzothiazole
B2344-25G
B2344-5G
TCB2344-25GEA
106.58
USD
TCB2344-25G
TCB2344-5G
Benzothiazol-2-ol ≥98.0%
Benzothiazol-2-ol
CAS Number: 934-34-9
MDL Number: MFCD00022868
Molecular Formula: C7H5NOS
Molecular Weight: 151.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 138
Other Product Names / Synonyms: 2-Benzothiazolol, 2-Hydroxybenzothiazole
Formula:
C₇H₅NOS MW: 151.18 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD00022868 CAS Number: 934-34-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...