Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromopiperonal ≥98.0%

Supplier: TCI America


B2332-5G
TCB2332-5GEA 49.93 USD InStock
TCB2332-5G
6-Bromopiperonal ≥98.0%
6-Bromopiperonal
CAS Number: 15930-53-7
MDL Number: MFCD00022952
Molecular Formula: C8H5BrO3
Molecular Weight: 229.03
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 130

Other Product Names / Synonyms: 6-Bromo-3,4-methylenedioxybenzaldehyde, 6-Bromo-1,3-benzodioxole-5-carboxaldehyde


Formula: C₈H₅BrO₃
MW: 229,03 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00022952
CAS Number: 15930-53-7
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty