N-(tert-Butoxycarbonyl)-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Boc-L-aspartic Acid
, Boc-Asp-OH
B2270-5G
TCB2270-5GEA
46
USD
TCB2270-5G
N-(tert-Butoxycarbonyl)-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-aspartic acid
CAS Number: 13726-67-5
MDL Number: MFCD00037279
Molecular Formula: C9H15NO6
Molecular Weight: 233.22
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -5 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-aspartic Acid, Boc-Asp-OH
Formula:
C₉H₁₅NO₆ MW: 233.22 g/mol |
MDL Number:
MFCD00037279 CAS Number: 13726-67-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...