Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromophthalic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America


B2257-1G B2257-5G B2257-25G
TCB2257-1GEA 13.52 USD InStock
TCB2257-1G TCB2257-5G TCB2257-25G
4-Bromophthalic acid ≥98.0% (by titrimetric analysis)
4-Bromophthalic acid
CAS Number: 6968-28-1
MDL Number: MFCD00043538
Molecular Formula: C8H5BrO4
Molecular Weight: 245.03
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 166


Formula: C₈H₅BrO₄
MW: 245,03 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00043538
CAS Number: 6968-28-1
EINECS: 230-183-4
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty