tert-Butylferrocene ≥97.0%

Supplier: TCI America

Synonyms: t-Butylferrocene

B2063-1G B2063-5G
TCB2063-005GEA 155.23 USD
TCB2063-005G TCB2063-001G
tert-Butylferrocene ≥97.0%
tert-Butylferrocene

CAS Number: 1316-98-9
MDL Number: MFCD00058729
Molecular Formula: C14H18Fe
Molecular Weight: 242.14
Purity/Analysis Method: 97.0% (GC)
Form: Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.18

https://us.vwr.com/stibo/web/std.lang.all/38/35/28933835.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR