Benzylmagnesium chloride ∼16% ∼1 mol/L in THF
B1933-250G
TCB1933-250GEA
280.83
USD
TCB1933-250G
Benzylmagnesium chloride ∼16% ∼1 mol/L in THF
Benzylmagnesium chloride
CAS Number: 6921-34-2
MDL Number: MFCD00000469
Molecular Formula: C7H7ClMg
Molecular Weight: 150.89
Flash Point (°C): -14
Formula:
C₇H₇ClMg Flash Pt: –14 °C |
MDL Number:
MFCD00000469 CAS Number: 6921-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...